Octadecyltriethoxysilane ≥85.0%

Supplier: TCI America

Synonyms: Triethoxy(octadecyl)silane

O0165-25G
TCO0165-025GEA 104.55 USD
TCO0165-025G
Octadecyltriethoxysilane ≥85.0%
Octadecyltriethoxysilane

CAS Number: 7399-00-0
MDL Number: MFCD00039875
Molecular Formula: C24H52O3Si
Molecular Weight: 416.76
Purity/Analysis Method: 85.0% (GC)
Form: Clear Liquid
Boiling point (°C): 169
Melting point (°C): 12
Flash Point (°C): 189
Specific Gravity (20/20): 0.87


Product Categories: Materials Chemistry, Silane Coupling Agents
Other Product Names / Synonyms: Triethoxyoctadecylsilane, Triethoxystearylsilane

Formula: C₂₄H₅₂O₃Si
MW: 416.76 g/mol
Boiling Pt: 169 °C
Melting Pt: 12 °C
Density: 0.87 (20 °C)
Flash Pt: 189 °C
MDL Number: MFCD00039875
CAS Number: 7399-00-0
EINECS: 230-995-9
https://us.vwr.com/stibo/web/std.lang.all/04/55/28990455.jpg

Order Now

Specification Test Results

Purity/Analysis Method 85.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR