n-Octane ≥98.5%
Supplier: TCI America
|
Danger
|
O0151-500ML
TCO0151-500MLEA
1552.81
USD
TCO0151-500ML
n-Octane ≥98.5%
n-Octane
[for HPLC Solvent]
CAS Number: 111-65-9
MDL Number: MFCD00009556
Molecular Formula: C8H18
Molecular Weight: 114.23
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 125
Melting point (°C): -57
Flash Point (°C): 14
Specific Gravity (20/20): 0.70
Formula:
H₃C(CH₂)₆CH₃ MW: 114.23 g/mol Boiling Pt: 125 °C Melting Pt: –57 °C Density: 0.70 (20 °C) Flash Pt: 14 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009556 CAS Number: 111-65-9 EINECS: 203-892-1 UN: 1262 ADR: 3,II Merck Index: 12,06847 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...