1,7-Octadiyne ≥98.0%
Supplier: TCI America
|
Danger
|
Synonyms:
Octa-1,7-diyne
O0147-5ML
O0147-25ML
TCO0147-005MLEA
59.11
USD
TCO0147-005ML
TCO0147-025ML
1,7-Octadiyne ≥98.0%
1,7-Octadiyne
CAS Number: 871-84-1
MDL Number: MFCD00008580
Molecular Formula: C8H10
Molecular Weight: 106.17
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 135
Flash Point (°C): 23
Specific Gravity (20/20): 0.82
Formula:
C₈H₁₀ MW: 106.17 g/mol Boiling Pt: 135 °C Melting Pt: –32 °C Density: 0.82 (20 °C) Flash Pt: 23 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00008580 CAS Number: 871-84-1 EINECS: 212-815-0 UN: 3295 ADR: 3,III |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...