Octamethylcyclotetrasiloxane ≥98.0%

Supplier: TCI America
Warning

Synonyms: Silicone fluid DC 244Dow Corning 244® fluidVolasil 244

O0142-25ML O0142-100ML O0142-500ML
TCO0142-100MLEA 88.26 USD
TCO0142-100ML TCO0142-500ML TCO0142-025ML
Octamethylcyclotetrasiloxane ≥98.0%
Octamethylcyclotetrasiloxane

CAS Number: 556-67-2
MDL Number: MFCD00003269
Molecular Formula: C8H24O4Si4
Molecular Weight: 296.62
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 175
Flash Point (°C): 54
Freezing point (°C): 18
Specific Gravity (20/20): 0.96


Other Product Names / Synonyms: Dimethylsiloxane Cyclic Tetramer

Formula: [-Si(CH₃)₂O-]₄
MW: 296.62 g/mol
Boiling Pt: 175 °C
Melting Pt: 18 °C
Density: 0.96 (20 °C)
Flash Pt: 54 °C
MDL Number: MFCD00003269
CAS Number: 556-67-2
EINECS: 209-136-7
UN: 1993
ADR: 3,III
Merck Index: 12,06843
https://us.vwr.com/stibo/web/std.lang.all/04/35/28990435.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR