3-Octyn-1-ol ≥98.0%

Supplier: TCI America

Synonyms: Oct-3-yn-1-ol

O0141-5ML
TCO0141-005MLEA 96 USD
TCO0141-005ML
3-Octyn-1-ol ≥98.0%
3-Octyn-1-ol

CAS Number: 14916-80-4
MDL Number: MFCD00039552
Molecular Formula: C8H14O
Molecular Weight: 126.20
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 95
Flash Point (°C): 110
Specific Gravity (20/20): 0.88

Formula: C₈H₁₄O
MW: 126.2 g/mol
Boiling Pt: 95 °C
Density: 0.88 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00039552
CAS Number: 14916-80-4
EINECS: 238-986-1

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR