3-Octyne ≥99.0%

Supplier: TCI America

O0126-5ML
TCO0126-005MLEA 119.72 USD
TCO0126-005ML
3-Octyne ≥99.0%
3-Octyne

CAS Number: 15232-76-5
MDL Number: MFCD00027276
Molecular Formula: C8H14
Molecular Weight: 110.20
Purity/Analysis Method: 99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 133
Flash Point (°C): 21
Specific Gravity (20/20): 0.75
Storage Temperature: 0-10°C


Other Product Names / Synonyms: Butylethylacetylene

Formula: C₈H₁₄
MW: 110.2 g/mol
Boiling Pt: 133 °C
Melting Pt: –103.9 °C
Density: 0.75 (20 °C)
Flash Pt: 21 °C
MDL Number: MFCD00027276
CAS Number: 15232-76-5
EINECS: 239-280-6
UN: 3295
ADR: 3,III

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR