2-Octyne ≥99.0%

Supplier: TCI America

O0125-5ML O0125-25ML
TCO0125-005MLEA 45.66 USD
TCO0125-005ML TCO0125-25ML
2-Octyne ≥99.0%
2-Octyne

CAS Number: 2809-67-8
MDL Number: MFCD00027302
Molecular Formula: C8H14
Molecular Weight: 110.20
Purity/Analysis Method: 99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 137
Flash Point (°C): 18
Specific Gravity (20/20): 0.76


Other Product Names / Synonyms: Amylmethylacetylene, Methylpentylacetylene

Formula: C₈H₁₄
MW: 110.2 g/mol
Boiling Pt: 137 °C
Density: 0.76 (20 °C)
Flash Pt: 18 °C
MDL Number: MFCD00027302
CAS Number: 2809-67-8
EINECS: 220-553-3
UN: 3295
ADR: 3,III
https://us.vwr.com/stibo/web/std.lang.all/04/23/28990423.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR