DL-Ornithine hydrochloride ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
2,5-Diaminopentanoic acid hydrochloride
, H-DL-Orn-OH·HCl
, DL-Ornithine monohydrochloride
CAS Number: 1069-31-4
MDL Number: MFCD00065398
Molecular Formula: C5H12N2O2
Molecular Weight: 168.62
Purity/Analysis Method:
Form: Crystal
Product Categories: Bioscience, non-Proteinogenic Amino Acids, Proteinogenic Amino Acids
Other Product Names / Synonyms: H-DL-Orn-OH.HCl, 2,5-Diaminopentanoic Acid Monohydrochloride, 2,5-Diaminovaleric Acid Monohydrochloride
Formula:
C₅H₁₃ClN₂O₂ MW: 168.62 g/mol |
MDL Number:
MFCD00065398 CAS Number: 1069-31-4 EINECS: 213-956-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...