1-Octanethiol ≥98.0%

Supplier: TCI America

Synonyms: 1-Mercaptooctane n-Octanethiol n-Octyl mercaptan

O0025-25ML O0025-500ML
TCO0025-500MLEA 57.83 USD
TCO0025-500ML TCO0025-25ML
1-Octanethiol ≥98.0%
1-Octanethiol

CAS Number: 111-88-6
MDL Number: MFCD00004912
Molecular Formula: C8H18S
Molecular Weight: 146.29
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 200
Flash Point (°C): 81
Specific Gravity (20/20): 0.84


Product Categories: Materials Chemistry, Self-Assembled Monolayer (SAM) Forming Agents, Derivatizing Reagents for Solubility Improvement
Other Product Names / Synonyms: n-Octyl Mercaptan

Formula: C₈H₁₈S
MW: 146.3 g/mol
Boiling Pt: 200 °C
Melting Pt: –49 °C
Density: 0.84 (20 °C)
Flash Pt: 81 °C
Storage Temperature: Ambient
MDL Number: MFCD00004912
CAS Number: 111-88-6
EINECS: 203-918-1
UN: 3082
ADR: 9,III
https://us.vwr.com/stibo/web/std.lang.all/03/38/28990338.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR