n-Octacosane ≥98.0%
Supplier: TCI America
Synonyms:
N-octacosane
O0002-25G
TCO0002-025GEA
79.56
USD
TCO0002-025G
n-Octacosane ≥98.0%
n-Octacosane
CAS Number: 630-02-4
MDL Number: MFCD00009355
Molecular Formula: C28H58
Molecular Weight: 394.77
Purity/Analysis Method:
Form: Crystal
Boiling point (°C): 286
Melting point (°C): 62
Flash Point (°C): 228
Formula:
H₃C(CH₂)₂₆CH₃ MW: 394.77 g/mol Boiling Pt: 286 °C Melting Pt: 62 °C Density: 0.8067 g/cm³ (20 °C) Flash Pt: 228 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009355 CAS Number: 630-02-4 EINECS: 211-125-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...