5-Nitroisoquinoline

Supplier: TCI America

N0640-5G N0640-25G
TCN0640-025GEA 200 USD
TCN0640-025G TCN0640-005G
5-Nitroisoquinoline
5-Nitroisoquinoline

CAS Number: 607-32-9
MDL Number: MFCD00006905
Molecular Formula: C9H6N2O2
Molecular Weight: 174.16
Form: Crystal
Color: Pale Yellow
Melting point (°C): 108

Formula: C₉H₆N₂O₂
Boiling Pt: 340 °C (1013 hPa)
Melting Pt: 108 °C
Storage Temperature: Ambient
MDL Number: MFCD00006905
CAS Number: 607-32-9
EINECS: 210-133-8
https://us.vwr.com/stibo/web/std.lang.all/98/41/28989841.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR