2-Nonynoic acid methyl ester ≥98.0%

Supplier: TCI America

Synonyms: Methyl 2-nonynoateMethyl non-2-ynoate

N0595-25ML
TCN0595-025MLEA 35.53 USD
TCN0595-025ML
2-Nonynoic acid methyl ester ≥98.0%
2-Nonynoic acid methyl ester

CAS Number: 111-80-8
MDL Number: MFCD00009547
Molecular Formula: C10H16O2
Molecular Weight: 168.24
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 121
Flash Point (°C): 104
Specific Gravity (20/20): 0.92


Other Product Names / Synonyms: 2-Nonynoic Acid Methyl Ester

Formula: C₁₀H₁₆O₂
MW: 168.24 g/mol
Boiling Pt: 121 °C
Density: 0.92 (20 °C)
Flash Pt: 104 °C
MDL Number: MFCD00009547
CAS Number: 111-80-8
EINECS: 203-909-2
https://us.vwr.com/stibo/web/std.lang.all/97/79/28989779.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR