5-Nitro-2-furoic acid ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
5-Nitrofuran-2-carboxylic acid
N0557-5G
TCN0557-005GEA
53
USD
TCN0557-005G
5-Nitro-2-furoic acid ≥98.0% (by titrimetric analysis)
5-Nitro-2-furoic acid
CAS Number: 645-12-5
MDL Number: MFCD00003240
Molecular Formula: C5H3NO5
Molecular Weight: 157.08
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 186
Other Product Names / Synonyms: 2-Carboxy-5-nitrofuran, 5-Nitro-2-furoic Acid
Formula:
C₅H₃NO₅ MW: 157.08 g/mol Melting Pt: 186 °C |
MDL Number:
MFCD00003240 CAS Number: 645-12-5 EINECS: 211-432-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...