trans-4-Nitrocinnamaldehyde ≥98.0%
Supplier: TCI America
Synonyms:
(E)-3-(4-Nitrophenyl)-2-propenal
, (E)-4-Nitrocinnamaldehyde
, (E)-3-(4-Nitrophenyl)acrylaldehyde
, trans-3-(4-Nitrophenyl)-2-propenal
N0541-5G
TCN0541-005GEA
153.83
USD
TCN0541-005G
trans-4-Nitrocinnamaldehyde ≥98.0%
trans-4-Nitrocinnamaldehyde
CAS Number: 1734-79-8
MDL Number: MFCD00007379
Molecular Formula: C9H7NO3
Molecular Weight: 177.16
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 141
Product Categories: Bioscience, Phenylpropanoids
Other Product Names / Synonyms: 3-(4-Nitrophenyl)acrylaldehyde, 3-(4-Nitrophenyl)-2-propenal
Formula:
C₉H₇NO₃ MW: 177.16 g/mol Melting Pt: 141 °C |
MDL Number:
MFCD00007379 CAS Number: 1734-79-8 EINECS: 217-076-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...