2-Nitrobenzaldehyde oxime ≥85.0%
Supplier: TCI America
Synonyms:
2-Nitrobenzaldoxime
N0486-25G
N0486-5G
TCN0486-025GEA
92.33
USD
TCN0486-025G
TCN0486-5G
2-Nitrobenzaldehyde oxime ≥85.0%
2-Nitrobenzaldehyde oxime
[Deprotecting Agent]
CAS Number: 6635-41-2
MDL Number: MFCD00007187
Molecular Formula: C7H6N2O3
Molecular Weight: 166.14
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 95
Formula:
C₇H₆N₂O₃ Melting Pt: 95 °C |
MDL Number:
MFCD00007187 CAS Number: 6635-41-2 EINECS: 229-634-8 UN: 2811 ADR: 6.1,III |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...