Ethyl-4-nitrobenzoate ≥98.0%

Supplier: TCI America

Synonyms: 4-Nitrobenzoic acid ethyl esterEthyl 4-nitrobenzoate

N0485-25G
TCN0485-025GEA 42.63 USD
TCN0485-025G
Ethyl-4-nitrobenzoate ≥98.0%
Ethyl 4-nitrobenzoate

CAS Number: 99-77-4
MDL Number: MFCD00007351
Molecular Formula: C9H9NO4
Molecular Weight: 195.17
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Boiling point (°C): 182
Melting point (°C): 57


Other Product Names / Synonyms: 4-Nitrobenzoic Acid Ethyl Ester

Formula: C₉H₉NO₄
MW: 195.17 g/mol
Boiling Pt: 182 °C
Melting Pt: 57 °C
Storage Temperature: Ambient
MDL Number: MFCD00007351
CAS Number: 99-77-4
EINECS: 202-786-2
https://us.vwr.com/stibo/web/std.lang.all/96/44/28989644.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR