1,8-Nonadiyne ≥97.0%

Supplier: TCI America

N0406-5ML
TCN0406-005MLEA 161.39 USD
TCN0406-005ML
1,8-Nonadiyne ≥97.0%
1,8-Nonadiyne

CAS Number: 2396-65-8
MDL Number: MFCD00008581
Molecular Formula: C9H12
Molecular Weight: 120.20
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 162
Flash Point (°C): 41
Specific Gravity (20/20): 0.83
Storage Temperature: 0-10°C

Formula: C₉H₁₂
MW: 120.19 g/mol
Boiling Pt: 162 °C
Melting Pt: –27.3 °C
Density: 0.83 (20 °C)
Flash Pt: 41 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008581
CAS Number: 2396-65-8
EINECS: 219-254-0
UN: 3295
ADR: 3,III

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR