3-Nonyn-1-ol ≥99.0%

Supplier: TCI America

N0390-5ML
TCN0390-005MLEA 86.24 USD
TCN0390-005ML
3-Nonyn-1-ol ≥99.0%
3-Nonyn-1-ol

CAS Number: 31333-13-8
MDL Number: MFCD00002958
Molecular Formula: C9H16O
Molecular Weight: 140.23
Purity/Analysis Method: 99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 131
Flash Point (°C): 93
Specific Gravity (20/20): 0.88


Other Product Names / Synonyms: (2-Hydroxyethyl)pentylacetylene

Formula: C₉H₁₆O
MW: 140.23 g/mol
Boiling Pt: 131 °C
Density: 0.88 (20 °C)
Flash Pt: 93 °C
MDL Number: MFCD00002958
CAS Number: 31333-13-8
EINECS: 250-573-8

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR