Dinonyl sulfide ≥97.0%

Supplier: TCI America

N0360-25G
TCN0360-25GEA 96.39 USD
TCN0360-25G
Dinonyl sulfide ≥97.0%
Dinonyl sulphide

CAS Number: 929-98-6
MDL Number: MFCD00027342
Molecular Formula: C18H38S
Molecular Weight: 286.56
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 202
Flash Point (°C): 110
Freezing point (°C): 15
Specific Gravity (20/20): 0.84


Other Product Names / Synonyms: Dinonyl Sulfide

Formula: C₁₈H₃₈S
MW: 286.57 g/mol
Boiling Pt: 202 °C
Melting Pt: 14.5 °C
Density: 0.84 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00027342
CAS Number: 929-98-6
UN: 3334
ADR: 9,
https://us.vwr.com/stibo/web/std.lang.all/94/43/28989443.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR