(2-Nitrophenyl)acetic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
2-(2-Nitrophenyl)acetic acid
N0353-25G
TCN0353-025GEA
57
USD
TCN0353-025G
(2-Nitrophenyl)acetic acid ≥98.0% (by GC, titration analysis)
(2-Nitrophenyl)acetic acid
CAS Number: 3740-52-1
MDL Number: MFCD00007190
Molecular Formula: C8H7NO4
Molecular Weight: 181.15
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 141
Formula:
(C₆H₄NO₂)CH₂COOH MW: 181.15 g/mol Melting Pt: 141 °C |
MDL Number:
MFCD00007190 CAS Number: 3740-52-1 EINECS: 223-128-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...