Norbornadiene ≥97.0% stabilized

Supplier: TCI America

Synonyms: 2,5-Norbornadiene

Stabilization: Stabilised with Stabilised

N0346-25ML N0346-500ML N0346-100ML
TCN0346-500MLEA 570 USD
TCN0346-500ML TCN0346-100ML TCN0346-25ML
Norbornadiene ≥97.0% stabilized
Norbornadiene

(stabilized with BHT)
CAS Number: 121-46-0
MDL Number: MFCD00082301
Molecular Formula: C7H8
Molecular Weight: 92.14
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 90
Flash Point (°C): -19
Specific Gravity (20/20): 0.91
Lambda max.: 195 nm (EtOH)


Product Categories: Synthetic Organic Chemistry, Ligands, Building Blocks ([2+2+2] Cycloaddition Reaction)
Other Product Names / Synonyms: Bicyclo[2.2.1]hepta-2,5-diene, NBD

Formula: C₇H₈
Boiling Pt: 90 °C
Density: 0.91 (20 °C)
Flash Pt: –19 °C
MDL Number: MFCD00082301
CAS Number: 121-46-0
EINECS: 204-472-0
https://us.vwr.com/stibo/web/std.lang.all/94/29/28989429.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR