DL-Norleucine ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: (±)-Norleucine H-DL-Nle-OH

N0302-25G
TCN0302-025GEA 78.16 USD
TCN0302-025G
DL-Norleucine ≥98.0% (by titrimetric analysis)
DL-Norleucine

CAS Number: 616-06-8
MDL Number: MFCD00064422
Molecular Formula: C6H13NO2
Molecular Weight: 131.18
Purity/Analysis Method: 98.0% (T)
Form: Crystal


Product Categories: Bioscience, non-Proteinogenic Amino Acids
Other Product Names / Synonyms: DL-2-Aminohexanoic Acid

Formula: C₆H₁₃NO₂
MW: 131.17 g/mol
Boiling Pt: 246 °C (1013 hPa)
Melting Pt: 113 °C
Density: 1.17 g/cm³ (20 °C)
MDL Number: MFCD00064422
CAS Number: 616-06-8
EINECS: 210-462-7
Merck Index: 12,06800
https://us.vwr.com/stibo/web/std.lang.all/93/80/28989380.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR