1-Nitronaphthalene ≥98.0%

Supplier: TCI America

Synonyms: α-Nitronaphthalene

N0212-25G N0212-500G
TCN0212-500GEA 122.82 USD
TCN0212-500G TCN0212-25G
1-Nitronaphthalene ≥98.0%
1-Nitronaphthalene

CAS Number: 86-57-7
MDL Number: MFCD00003913
Molecular Formula: C10H7NO2
Molecular Weight: 173.17
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 58
Flash Point (°C): 163

Formula: C₁₀H₇NO₂
MW: 173.17 g/mol
Boiling Pt: 304 °C (1013 hPa)
Melting Pt: 58 °C
Density: 1.331 g/cm³ (20 °C)
Flash Pt: 163 °C
Storage Temperature: Ambient
MDL Number: MFCD00003913
CAS Number: 86-57-7
EINECS: 201-684-5
UN: 2538
ADR: 4.1,III
Merck Index: 12,06711
https://us.vwr.com/stibo/web/std.lang.all/93/02/28989302.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR