Nitromethane ≥98.0%
Supplier: TCI America
|
Danger
|
N0209-25G
N0209-100G
N0209-500G
TCN0209-500GEA
110.58
USD
TCN0209-500G
TCN0209-100G
TCN0209-25G
Nitromethane ≥98.0%
Nitromethane
CAS Number: 75-52-5
MDL Number: MFCD00007400
Molecular Formula: CH3NO2
Molecular Weight: 61.04
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 101
Melting point (°C): -29
Flash Point (°C): 35
Specific Gravity (20/20): 1.14
Lambda max.: 380 nm (H2O)
Formula:
CH₃NO₂ MW: 61.04 g/mol Boiling Pt: 101 °C Melting Pt: –29 °C Density: 1.14 (20 °C) Flash Pt: 35 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007400 CAS Number: 75-52-5 EINECS: 200-876-6 UN: 1261 ADR: 3,II Merck Index: 12,06708 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...