2-Nitroanisole ≥98.0%
Supplier: TCI America
|
Danger
|
N0125-500G
N0125-25G
TCN0125-500GEA
60.29
USD
TCN0125-500G
TCN0125-25G
2-Nitroanisole ≥98.0%
2-Nitroanisole
CAS Number: 91-23-6
MDL Number: MFCD00007096
Molecular Formula: C7H7NO3
Molecular Weight: 153.14
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 273
Flash Point (°C): 124
Freezing point (°C): 9
Specific Gravity (20/20): 1.25
Other Product Names / Synonyms: 1-Methoxy-2-nitrobenzene
Formula:
C₇H₇NO₃ MW: 153.14 g/mol Boiling Pt: 273 °C Melting Pt: 9 °C Density: 1.25 (20 °C) Flash Pt: 124 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007096 CAS Number: 91-23-6 EINECS: 202-052-1 UN: 2730 ADR: 6.1,III Merck Index: 12,06682 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...