2-Nitroaniline ≥99.0%

Supplier: TCI America
Danger

Synonyms: o-Nitroaniline

N0118-25G N0118-500G
TCN0118-25GEA 27.91 USD
TCN0118-25G TCN0118-500G
2-Nitroaniline ≥99.0%
2-Nitroaniline

CAS Number: 88-74-4
MDL Number: MFCD00007687
Molecular Formula: C6H6N2O2
Molecular Weight: 138.13
Purity/Analysis Method: 99.0% (GC)
Form: Crystal
Boiling point (°C): 165
Melting point (°C): 73
Flash Point (°C): 168

Formula: C₆H₆N₂O₂
MW: 138.13 g/mol
Boiling Pt: 165 °C
Melting Pt: 73 °C
Density: 1.446 g/cm³ (20 °C)
Flash Pt: 168 °C
Storage Temperature: Ambient
MDL Number: MFCD00007687
CAS Number: 88-74-4
EINECS: 201-855-4
UN: 1661
ADR: 6.1,II
Merck Index: 12,06680
https://us.vwr.com/stibo/web/std.lang.all/92/15/28989215.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR