2-Nitro-p-toluidine ≥98.0%

Supplier: TCI America
Danger

Synonyms: 4-Methyl-2-nitroaniline4-Amino-3-nitrotoluene2-Nitro-4-methylaniline

N0114-25G N0114-500G
TCN0114-500GEA 227.55 USD
TCN0114-500G TCN0114-25G
2-Nitro-p-toluidine ≥98.0%
2-Nitro-p-toluidine

CAS Number: 89-62-3
MDL Number: MFCD00007907
Molecular Formula: C7H8N2O2
Molecular Weight: 152.15
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 116
Flash Point (°C): 157


Other Product Names / Synonyms: 4-Amino-3-nitrotoluene, 2-Nitro-p-toluidine

Formula: C₇H₈N₂O₂
MW: 152.15 g/mol
Boiling Pt: 303 °C (1013 hPa)
Melting Pt: 116 °C
Density: 1.164 g/cm³ (20 °C)
Flash Pt: 157 °C
Storage Temperature: Ambient
MDL Number: MFCD00007907
CAS Number: 89-62-3
EINECS: 201-924-9
UN: 2660
ADR: 6.1,III
https://us.vwr.com/stibo/web/std.lang.all/92/11/28989211.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR