(1-Naphthoxy)acetic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 2-(Naphthalen-1-yloxy)acetic acid

N0046-10G
TCN0046-010GEA 450 USD
TCN0046-010G
(1-Naphthoxy)acetic acid ≥98.0% (by titrimetric analysis)
(1-Naphthoxy)acetic acid

CAS Number: 2976-75-2
MDL Number: MFCD00003927
Molecular Formula: C12H10O3
Molecular Weight: 202.21
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Melting point (°C): 196

Formula: C₁₂H₁₀O₃
MW: 202.21 g/mol
Melting Pt: 196 °C
MDL Number: MFCD00003927
CAS Number: 2976-75-2
EINECS: 221-023-4
https://us.vwr.com/stibo/web/std.lang.all/91/64/28989164.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR