1-Naphthoic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

Synonyms: α-Naphthoic acid

N0024-500G N0024-25G
TCN0024-500GEA 251.97 USD
TCN0024-500G TCN0024-025G
1-Naphthoic acid ≥98.0% (by titrimetric analysis)
1-Naphthoic acid

CAS Number: 86-55-5
MDL Number: MFCD00004007
Molecular Formula: C11H8O2
Molecular Weight: 172.18
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Melting point (°C): 164


Other Product Names / Synonyms: 1-Naphthalenecarboxylic Acid

Formula: C₁₁H₈O₂
MW: 172.18 g/mol
Boiling Pt: 300 °C (1013 hPa)
Melting Pt: 164 °C
Density: 1 g/cm³ (20 °C)
MDL Number: MFCD00004007
CAS Number: 86-55-5
EINECS: 201-681-9
Merck Index: 12,06468
https://us.vwr.com/stibo/web/std.lang.all/91/45/28989145.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR