3-Methyl-2-nitrobenzoic acid ≥98.0% (by titrimetric analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
3-Methyl-2-nitrobenzoic acid
, 2-Nitro-m-toluic acid
M1370-250G
M1370-25G
TCM1370-25GEA
63.95
USD
TCM1370-25G
TCM1370-250G
3-Methyl-2-nitrobenzoic acid ≥98.0% (by titrimetric analysis)
2-Nitro-m-toluic acid
Form: Crystal
Melting point (°C): 223
Formula:
C₈H₇NO₄ MW: 181.15 g/mol Boiling Pt: 340 °C (1013 hPa) Melting Pt: 223 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007180 CAS Number: 5437-38-7 EINECS: 226-610-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...