2-Methyl-3-nitrobenzotrifluoride ≥97.0%

M1275-5G M1275-25G
TCM1275-005GEA 106.09 USD
TCM1275-005G TCM1275-025G
2-Methyl-3-nitrobenzotrifluoride ≥97.0%
2-Methyl-3-nitrobenzotrifluoride

CAS Number: 6656-49-1
MDL Number: MFCD00042322
Molecular Formula: C8H6F3NO2
Molecular Weight: 205.14
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.40


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 2-Nitro-6-(trifluoromethyl)toluene

Formula: C₈H₆F₃NO₂
MW: 205.14 g/mol
Boiling Pt: 35…38 °C (0.04 mmHg)
Density: 1.40 (20 °C)
MDL Number: MFCD00042322
CAS Number: 6656-49-1
EINECS: 229-688-2
UN: 2810
ADR: 6.1,III
https://us.vwr.com/stibo/web/std.lang.all/51/11/28975111.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR