4-Methylbenzhydrol ≥98.0%

Supplier: TCI America

M1176-25G
TCM1176-025GEA 109 USD
TCM1176-025G
4-Methylbenzhydrol ≥98.0%
4-Methylbenzhydrol

CAS Number: 1517-63-1
MDL Number: MFCD00004492
Molecular Formula: C14H14O
Molecular Weight: 198.27
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 54
Flash Point (°C): 113


Other Product Names / Synonyms: 4-Methyldiphenylmethanol

Formula: C₁₄H₁₄O
Melting Pt: 54 °C
Flash Pt: 113 °C
MDL Number: MFCD00004492
CAS Number: 1517-63-1
EINECS: 216-171-1
https://us.vwr.com/stibo/web/std.lang.all/49/09/28974909.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR