3-Methylbenzyl alcohol ≥98.0%
Supplier: TCI America
Synonyms:
3-(Hydroxymethyl)toluole
, (3-Methylphenyl)methanol
M1124-25G
TCM1124-025GEA
87.29
USD
TCM1124-025G
3-Methylbenzyl alcohol ≥98.0%
3-Methylbenzyl alcohol
CAS Number: 587-03-1
MDL Number: MFCD00004646
Molecular Formula: C8H10O
Molecular Weight: 122.17
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 217
Flash Point (°C): 105
Specific Gravity (20/20): 1.02
Other Product Names / Synonyms: alpha-Hydroxy-m-xylene
Formula:
C₈H₁₀O MW: 122.17 g/mol Boiling Pt: 217 °C Melting Pt: < –20 °C Density: 1.02 (20 °C) Flash Pt: 105 °C |
MDL Number:
MFCD00004646 CAS Number: 587-03-1 EINECS: 209-595-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...