N-Methylmaleamic acid ≥98.0% (by titrimetric analysis)
Supplier: TCI America
M1000-25G
TCM1000-025GEA
99
USD
TCM1000-025G
N-Methylmaleamic acid ≥98.0% (by titrimetric analysis)
N-Methylmaleamic acid
CAS Number: 6936-48-7
MDL Number: MFCD00014012
Molecular Formula: C5H7NO3
Molecular Weight: 129.12
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 150
Other Product Names / Synonyms: 3-(Methylaminocarbonyl)acrylic Acid
Formula:
C₅H₇NO₃ Melting Pt: 150 °C |
MDL Number:
MFCD00014012 CAS Number: 6936-48-7 EINECS: 230-062-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...