(±)-3-Methyl-3-heptanol

Supplier: TCI America

M0955-5ML
TCM0955-005MLEA 243 USD
TCM0955-005ML
(±)-3-Methyl-3-heptanol
(±)-3-Methyl-3-heptanol

CAS Number: 5582-82-1
MDL Number: MFCD00021841
Molecular Formula: C8H18O
Molecular Weight: 130.23
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 64
Flash Point (°C): 58
Specific Gravity (20/20): 0.83


Other Product Names / Synonyms: Butylethylmethylcarbinol

Formula: C₈H₁₈O
Boiling Pt: 64 °C
Density: 0.83 (20 °C)
Flash Pt: 58 °C
MDL Number: MFCD00021841
CAS Number: 5582-82-1
UN: 1987
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR