2-Methoxyfuran ≥98.0%

M0936-5G
TCM0936-005GEA 212.01 USD
TCM0936-005G
2-Methoxyfuran ≥98.0%
2-Methoxyfuran

CAS Number: 25414-22-6
MDL Number: MFCD00003227
Molecular Formula: C5H6O2
Molecular Weight: 98.10
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 110
Flash Point (°C): 10
Specific Gravity (20/20): 1.05
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 2-Furyl Methyl Ether

Formula: C₅H₆O₂
Boiling Pt: 110 °C
Density: 1.05 (20 °C)
Flash Pt: 10 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00003227
CAS Number: 25414-22-6
EINECS: 246-953-8
UN: 3271
ADR: 3,II

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR