1-Methyl-2-piperidinemethanol ≥98.0% (by GC, titration analysis)
Supplier: TCI America
M0923-25ML
TCM0923-025MLEA
289.78
USD
TCM0923-025ML
1-Methyl-2-piperidinemethanol ≥98.0% (by GC, titration analysis)
1-Methyl-2-piperidinemethanol
CAS Number: 20845-34-5
MDL Number: MFCD00006494
Molecular Formula: C7H15NO
Molecular Weight: 129.20
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 98
Flash Point (°C): 81
Specific Gravity (20/20): 0.98
Other Product Names / Synonyms: 2-Hydroxymethyl-1-methylpiperidine
Formula:
C₇H₁₅NO Boiling Pt: 98 °C Density: 0.98 (20 °C) Flash Pt: 81 °C |
MDL Number:
MFCD00006494 CAS Number: 20845-34-5 EINECS: 244-078-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...