2-Methylbenzophenone ≥98.0%

Supplier: TCI America

Synonyms: o-Methylbenzophenone

M0903-25G
TCM0903-025GEA 67.66 USD
TCM0903-025G
2-Methylbenzophenone ≥98.0%
2-Methylbenzophenone

CAS Number: 131-58-8
MDL Number: MFCD00008518
Molecular Formula: C14H12O
Molecular Weight: 196.25
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 168
Melting point (°C): -18
Flash Point (°C): 113
Specific Gravity (20/20): 1.09

Formula: C₁₄H₁₂O
MW: 196.25 g/mol
Boiling Pt: 168 °C
Melting Pt: –18 °C
Density: 1.09 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00008518
CAS Number: 131-58-8
EINECS: 205-032-0
Merck Index: 12,07470
https://us.vwr.com/stibo/web/std.lang.all/44/33/28974433.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR