2-(4-Methoxyphenyl)ethanol ≥98.0%

Supplier: TCI America

Synonyms: 4-Methoxyphenethyl alcohol

M0889-25G
TCM0889-025GEA 93.38 USD
TCM0889-025G
2-(4-Methoxyphenyl)ethanol ≥98.0%
2-(4-Methoxyphenyl)ethanol

CAS Number: 702-23-8
MDL Number: MFCD00002900
Molecular Formula: C9H12O2
Molecular Weight: 152.19
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Boiling point (°C): 336
Flash Point (°C): 113
Freezing point (°C): 27


Other Product Names / Synonyms: 4-(2-Hydroxyethyl)anisole, 2-(4-Methoxyphenyl)ethanol

Formula: C₉H₁₂O₂
MW: 152.19 g/mol
Boiling Pt: 336 °C
Flash Pt: 113 °C
MDL Number: MFCD00002900
CAS Number: 702-23-8
EINECS: 211-866-6
https://us.vwr.com/stibo/web/std.lang.all/44/04/28974404.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR