3-Methylpyridazine ≥97.0%

Supplier: TCI America
Warning

M0872-5ML M0872-25ML
TCM0872-025MLEA 853.62 USD
TCM0872-025ML TCM0872-005ML
3-Methylpyridazine ≥97.0%
3-Methylpyridazine

CAS Number: 1632-76-4
MDL Number: MFCD00006469
Molecular Formula: C5H6N2
Molecular Weight: 94.12
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 215
Flash Point (°C): 87
Specific Gravity (20/20): 1.05

Formula: C₅H₆N₂
MW: 94.12 g/mol
Boiling Pt: 215 °C
Density: 1.05 (20 °C)
Flash Pt: 87 °C
MDL Number: MFCD00006469
CAS Number: 1632-76-4
EINECS: 216-640-0
https://us.vwr.com/stibo/web/std.lang.all/43/77/28974377.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR