1-Methyl-3-piperidinemethanol ≥95.0%
Supplier: TCI America
Synonyms:
3-(Hydroxymethyl)-1-methylpiperidine
M0859-25ML
TCM0859-025MLEA
83.23
USD
TCM0859-025ML
1-Methyl-3-piperidinemethanol ≥95.0%
1-Methyl-3-piperidinemethanol
CAS Number: 7583-53-1
MDL Number: MFCD00006497
Molecular Formula: C7H15NO
Molecular Weight: 129.20
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 114
Flash Point (°C): 94
Specific Gravity (20/20): 0.98
Other Product Names / Synonyms: 3-Hydroxymethyl-1-methylpiperidine
Formula:
C₇H₁₅NO MW: 129.2 g/mol Boiling Pt: 114 °C Density: 0.98 (20 °C) Flash Pt: 94 °C |
MDL Number:
MFCD00006497 CAS Number: 7583-53-1 EINECS: 231-488-5 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...