Pyridine-2-aldoxime methochloride ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
2-PAM chloride
, 2-((Hydroxyimino)methyl)-1-methylpyridin-1-ium chloride
, Pralidoxime Chloride
, Pyridine-2-carboxaldoxime methochloride
M0819-25G
TCM0819-025GEA
105.56
USD
TCM0819-025G
Pyridine-2-aldoxime methochloride ≥98.0% (by HPLC, titration analysis)
Pyridine-2-aldoxime methochloride
CAS Number: 51-15-0
MDL Number: MFCD00011981
Molecular Formula: C7H9ClN2O
Molecular Weight: 172.61
Purity/Analysis Method:
Form: Crystal
Other Product Names / Synonyms: Pralidoxime Chloride
Formula:
C₇H₉ClN₂O MW: 172.61 g/mol |
MDL Number:
MFCD00011981 CAS Number: 51-15-0 EINECS: 200-080-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...