N-Methyl-2,2,2-trifluoroacetamide ≥98.0%

Supplier: TCI America

Synonyms: N-Methyltrifluoroacetamide2,2,2-Trifluoro-N-methylacetamide

M0817-25G
TCM0817-025GEA 29.44 USD
TCM0817-025G
N-Methyl-2,2,2-trifluoroacetamide ≥98.0%
N-Methyl-2,2,2-trifluoroacetamide

CAS Number: 815-06-5
MDL Number: MFCD00009670
Molecular Formula: C3H4F3NO
Molecular Weight: 127.07
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Boiling point (°C): 160
Melting point (°C): 52
Flash Point (°C): 73

Formula: C₃H₄F₃NO
Boiling Pt: 160 °C
Melting Pt: 52 °C
Flash Pt: 73 °C
MDL Number: MFCD00009670
CAS Number: 815-06-5
EINECS: 212-417-7
https://us.vwr.com/stibo/web/std.lang.all/42/87/28974287.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR