4-(4-Methoxyphenyl)-2-butanone ≥97.0%
Supplier: TCI America
|
Danger
|
Synonyms:
1-(4-Methoxyphenyl)-3-butanone
M0756-25ML
TCM0756-025MLEA
53.8
USD
TCM0756-025ML
4-(4-Methoxyphenyl)-2-butanone ≥97.0%
4-(4-Methoxyphenyl)-2-butanone
CAS Number: 104-20-1
MDL Number: MFCD00008791
Molecular Formula: C11H14O2
Molecular Weight: 178.23
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 177
Melting point (°C): 10
Flash Point (°C): 100
Specific Gravity (20/20): 1.05
Other Product Names / Synonyms: Anisylacetone, 4-Methoxybenzylacetone
Formula:
C₁₁H₁₄O₂ MW: 178.23 g/mol Boiling Pt: 177 °C Melting Pt: 10 °C Density: 1.05 (20 °C) Flash Pt: 100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008791 CAS Number: 104-20-1 EINECS: 203-184-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...