4-Methylvaleric acid ≥97.0%
Supplier: TCI America
|
Danger
|
Synonyms:
Isocaproic acid
, 4-Methylpentanoic acid
M0750-25ML
TCM0750-25MLEA
90.44
USD
TCM0750-25ML
4-Methylvaleric acid ≥97.0%
4-Methylvaleric acid
CAS Number: 646-07-1
MDL Number: MFCD00002803
Molecular Formula: C6H12O2
Molecular Weight: 116.16
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 201
Flash Point (°C): 97
Specific Gravity (20/20): 0.92
Other Product Names / Synonyms: 4-Methylpentanoic Acid
Formula:
C₆H₁₂O₂ MW: 116.16 g/mol Boiling Pt: 201 °C Melting Pt: –35 °C Density: 0.92 (20 °C) Flash Pt: 97 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002803 CAS Number: 646-07-1 EINECS: 211-464-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...