Tetradecanenitrile ≥96.0%

Supplier: TCI America

Synonyms: Tetradecanonitrile

M0740-25ML M0740-500ML
TCM0740-500MLEA 410 USD
TCM0740-500ML TCM0740-025ML
Tetradecanenitrile ≥96.0%
Tetradecanenitrile

CAS Number: 629-63-0
MDL Number: MFCD00019848
Molecular Formula: C14H27N
Molecular Weight: 209.38
Purity/Analysis Method: 96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 227
Specific Gravity (20/20): 0.83


Other Product Names / Synonyms: Tetradecanonitrile, Tridecyl Cyanide

Formula: C₁₄H₂₇N
Boiling Pt: 227 °C
Density: 0.83 (20 °C)
MDL Number: MFCD00019848
CAS Number: 629-63-0
EINECS: 211-099-7
https://us.vwr.com/stibo/web/std.lang.all/42/16/28974216.jpg

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR