N-Methylisobutylamine ≥98.0%

Supplier: TCI America

M0691-25ML
TCM0691-025MLEA 146 USD
TCM0691-025ML
N-Methylisobutylamine ≥98.0%
N-Methylisobutylamine

CAS Number: 625-43-4
MDL Number: MFCD00015043
Molecular Formula: C5H13N
Molecular Weight: 87.17
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 78
Specific Gravity (20/20): 0.73


Other Product Names / Synonyms: N-Isobutylmethylamine

Formula: C₅H₁₃N
Boiling Pt: 78 °C
Density: 0.73 (20 °C)
MDL Number: MFCD00015043
CAS Number: 625-43-4
EINECS: 210-893-0
https://us.vwr.com/stibo/web/std.lang.all/41/66/28974166.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR