4-Methoxyphenylhydrazinium chloride ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 4-Methoxyphenylhydrazine hydrochloride

M0637-5G M0637-25G
TCM0637-025GEA 190.91 USD
TCM0637-025G TCM0637-5G
4-Methoxyphenylhydrazinium chloride ≥98.0% (by HPLC, titration analysis)
4-Methoxyphenylhydrazinium chloride

CAS Number: 19501-58-7
MDL Number: MFCD00012945
Molecular Formula: C7H10N2O
Molecular Weight: 174.63
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal


Other Product Names / Synonyms: 4-Hydrazinoanisole Hydrochloride

Formula: C₇H₁₁ClN₂O
MW: 174.63 g/mol
MDL Number: MFCD00012945
CAS Number: 19501-58-7
EINECS: 243-115-3
https://us.vwr.com/stibo/web/std.lang.all/41/00/28974100.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR