Methylcyclohexane ≥99.0%

Supplier: TCI America
Danger

M0627-500ML
TCM0627-500MLEA 128 USD
TCM0627-500ML
Methylcyclohexane ≥99.0%
Methylcyclohexane

[for HPLC Solvent]
CAS Number: 108-87-2
MDL Number: MFCD00001497
Molecular Formula: C7H14
Molecular Weight: 98.19
Purity/Analysis Method: 99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 100
Melting point (°C): -127
Flash Point (°C): -4
Specific Gravity (20/20): 0.77


Product Categories: Analytical Science, Solvents for HPLC
Other Product Names / Synonyms: Hexahydrotoluene, MCH

Formula: C₆H₁₁CH₃
MW: 98.19 g/mol
Boiling Pt: 100 °C
Melting Pt: –127 °C
Density: 0.77 (20 °C)
Flash Pt: –4 °C
Storage Temperature: Ambient
MDL Number: MFCD00001497
CAS Number: 108-87-2
EINECS: 203-624-3
UN: 2296
ADR: 3,II
Merck Index: 12,10412
https://us.vwr.com/stibo/web/std.lang.all/40/87/28974087.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR