(±)-3-Methyl-1-pentene ≥98.0%

Supplier: TCI America

M0575-1G M0575-5G
TCM0575-005GEA 173.57 USD
TCM0575-005G TCM0575-1G
(±)-3-Methyl-1-pentene ≥98.0%
(±)-3-Methyl-1-pentene

CAS Number: 760-20-3
MDL Number: MFCD00009339
Molecular Formula: C6H12
Molecular Weight: 84.16
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 54
Flash Point (°C): -30
Specific Gravity (20/20): 0.67


Other Product Names / Synonyms: 2-Vinylbutane

Formula: C₆H₁₂
MW: 84.16 g/mol
Boiling Pt: 54 °C
Melting Pt: –153 °C
Density: 0.67 (20 °C)
Flash Pt: –30 °C
Storage Temperature: Ambient
MDL Number: MFCD00009339
CAS Number: 760-20-3
EINECS: 212-076-4
UN: 3295
ADR: 3,II
https://us.vwr.com/stibo/web/std.lang.all/40/33/28974033.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR